siRNA Details
Gene Symbol: CDHR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171968.1 | NP_001165439.1 | 11 | 1380 | ||
NM_021924.4 | NP_068743.2 | 11 | 1380 | |||
NM_031264.3 | NP_112554.2 | 11 | 1380 | |||
XM_006718253.3 | XP_006718316.1 | 11 | 1280 | |||
XM_011520188.2 | XP_011518490.1 | 11 | 1280 | |||
XM_011520189.2 | XP_011518491.1 | 11 | 1280 | |||
XM_011520190.2 | XP_011518492.1 | 11 | 1280 | |||
XM_011520191.2 | XP_011518493.1 | 11 | 1280 | |||
GenBank mRNA | AF258675.1 | - | 12 | 1536 | ||
AF258676.1 | - | 12 | 1536 | |||
AF276242.1 | - | 11 | 1215 | |||
AF301909.1 | - | 11 | 1215 | |||
AK000226.1 | - | 11 | 1398 | |||
AK000384.1 | - | 11 | 1285 | |||
AK025012.1 | - | 4 | 367 | |||
AK300991.1 | - | 12 | 1232 | |||
AK308040.1 | - | 11 | 1304 | |||
AK309157.1 | - | 10 | 1175 | |||
AY358368.1 | - | 11 | 1237 | |||
BC038405.1 | - | 11 | 1237 | 5185584 | ||
DQ491193.1 | - | 11 | 1287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
