siRNA Details
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138410.3 | NP_612419.1 | 5 | 1049 | ||
NM_181472.2 | NP_852137.1 | 4 | 950 | |||
GenBank mRNA | AF479263.2 | - | 5 | 1049 | ||
AK055554.1 | - | 2 | 1770 | |||
AK308878.1 | - | 4 | 648 | |||
BC010116.2 | - | 5 | 831 | 3636045 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
