siRNA Details
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138410.3 | NP_612419.1 | 1, 2 | 390 | ||
NM_181472.2 | NP_852137.1 | 1, 2 | 390 | |||
XM_011533319.2 | XP_011531621.1 | 1, 2 | 216 | |||
XM_017005646.1 | XP_016861135.1 | 1, 2 | 236 | |||
GenBank mRNA | AF479263.2 | - | 1, 2 | 390 | ||
AK308878.1 | - | 1, 2 | 88 | |||
AY174113.1 | - | 1, 2 | 154 | |||
BC010116.2 | - | 1, 2 | 172 | 3636045 | ||
DQ132881.1 | - | 1, 2 | 154 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
