siRNA Details
Gene Symbol: MAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002371.3 | NP_002362.1 | 4 | 1009 | ||
NM_022438.2 | NP_071883.1 | 3 | 883 | |||
NM_022439.2 | NP_071884.1 | 3 | 841 | |||
NM_022440.2 | NP_071885.1 | 2 | 715 | |||
GenBank mRNA | AK123269.1 | - | 1 | 2212 | ||
BC000458.2 | - | 4 | 980 | 2822079 | ||
BC003006.1 | - | 4 | 980 | 2822079 | ||
M15800.1 | - | 4 | 925 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
