siRNA Details
Gene Symbol: TRIM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032546.3 | NP_115935.3 | 8 | 1370 | ||
NM_187841.2 | NP_912730.2 | 7 | 1244 | |||
GenBank mRNA | AJ291714.1 | - | 8 | 1067 | ||
AK300699.1 | - | 8 | 1216 | |||
BC141807.1 | - | 7 | 1016 | 40147443 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
