siRNA Details
Gene Symbol: EIF4G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194946.1 | NP_001181875.1 | 23 | 3542 | ||
NM_001194947.1 | NP_001181876.1 | 22 | 3485 | |||
NM_001291157.1 | NP_001278086.1 | 22 | 3574 | |||
NM_004953.4 | NP_004944.3 | 15 | 3033 | |||
NM_182917.4 | NP_886553.3 | 21 | 3669 | |||
NM_198241.2 | NP_937884.1 | 22 | 3521 | |||
NM_198242.2 | NP_937885.1 | 18 | 3106 | |||
NM_198244.2 | NP_937887.1 | 19 | 3283 | |||
GenBank mRNA | AB210013.1 | - | 21 | 3660 | ||
AF104913.1 | - | 19 | 3145 | |||
AF281070.1 | - | 21 | 3527 | |||
AK226160.1 | - | 22 | 3455 | |||
AK294567.1 | - | 20 | 3152 | |||
AK299763.1 | - | 19 | 3211 | |||
AL832754.1 | - | 18 | 2995 | |||
AY082886.1 | - | 21 | 3527 | |||
BC007788.2 | - | 4 | 389 | |||
BC082764.1 | - | 15 | 2679 | |||
BC140892.1 | - | 23 | 3384 | 9021760 | ||
BC140896.1 | - | 22 | 3327 | 9021764 | ||
BT007282.1 | - | 4 | 388 | |||
BX647812.1 | - | 22 | 3434 | |||
D12686.1 | - | 15 | 3024 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
