siRNA Details
Gene Symbol: EIF4G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194946.1 | NP_001181875.1 | 8, 9 | 821 | ||
NM_001194947.1 | NP_001181876.1 | 7, 8 | 764 | |||
NM_001291157.1 | NP_001278086.1 | 7, 8 | 853 | |||
NM_182917.4 | NP_886553.3 | 6, 7 | 945 | |||
NM_198241.2 | NP_937884.1 | 7, 8 | 800 | |||
NM_198242.2 | NP_937885.1 | 3, 4 | 385 | |||
NM_198244.2 | NP_937887.1 | 4, 5 | 562 | |||
GenBank mRNA | AB210013.1 | - | 6, 7 | 939 | ||
AF002815.1 | - | 4, 5 | 601 | |||
AF002816.1 | - | 4, 5 | 442 | |||
AF104913.1 | - | 4, 5 | 421 | |||
AF281070.1 | - | 6, 7 | 803 | |||
AK096719.1 | - | 5, 6 | 450 | |||
AK128378.1 | - | 6, 7 | 549 | |||
AK226160.1 | - | 7, 8 | 734 | |||
AK294567.1 | - | 5, 6 | 431 | |||
AK299763.1 | - | 4, 5 | 490 | |||
AK308319.1 | - | 2, 3 | 150 | |||
AL832754.1 | - | 3, 4 | 274 | |||
AY082886.1 | - | 6, 7 | 803 | |||
BC140892.1 | - | 8, 9 | 663 | 9021760 | ||
BC140896.1 | - | 7, 8 | 606 | 9021764 | ||
BU176883.1 | - | 3, 4 | 328 | |||
BX647812.1 | - | 7, 8 | 713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
