siRNA Details
Gene Symbol: EIF4G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194946.1 | NP_001181875.1 | 27 | 4130 | ||
NM_001194947.1 | NP_001181876.1 | 26 | 4073 | |||
NM_001291157.1 | NP_001278086.1 | 26 | 4162 | |||
NM_004953.4 | NP_004944.3 | 19 | 3621 | |||
NM_182917.4 | NP_886553.3 | 25 | 4257 | |||
NM_198241.2 | NP_937884.1 | 26 | 4109 | |||
NM_198242.2 | NP_937885.1 | 22 | 3694 | |||
NM_198244.2 | NP_937887.1 | 23 | 3871 | |||
GenBank mRNA | AB210013.1 | - | 25 | 4248 | ||
AF104913.1 | - | 23 | 3733 | |||
AF281070.1 | - | 25 | 4115 | |||
AK131407.1 | - | 6 | 808 | |||
AK226160.1 | - | 26 | 4043 | |||
AK294567.1 | - | 24 | 3740 | |||
AK299763.1 | - | 23 | 3799 | |||
AL832754.1 | - | 22 | 3583 | |||
AY082886.1 | - | 25 | 4115 | |||
BC007788.2 | - | 8 | 977 | |||
BC082764.1 | - | 19 | 3267 | |||
BC140892.1 | - | 27 | 3972 | 9021760 | ||
BC140896.1 | - | 26 | 3915 | 9021764 | ||
BT007282.1 | - | 8 | 976 | |||
BX647812.1 | - | 26 | 4022 | |||
CX165113.1 | - | 5 | 562 | |||
D12686.1 | - | 19 | 3612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
