siRNA Details
Gene Symbol: TAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003182.2 | NP_003173.1 | 3 | 392 | ||
NM_013996.2 | NP_054702.1 | 3 | 392 | |||
NM_013997.2 | NP_054703.1 | 3 | 392 | |||
NM_013998.2 | NP_054704.1 | 3 | 392 | |||
GenBank mRNA | AF050655.1 | - | 2 | 41 | ||
AF050656.1 | - | 2 | 41 | |||
AF050657.1 | - | 2 | 41 | |||
AF050658.1 | - | 2 | 41 | |||
AK222957.1 | - | 3 | 267 | |||
BC018047.1 | - | 3 | 266 | 4792746 | ||
CR407602.1 | - | 2 | 146 | |||
CR541730.1 | - | 2 | 146 | |||
DN992454.1 | - | 3 | 400 | |||
M68906.1 | - | 2 | 42 | |||
M68907.1 | - | 2 | 42 | |||
U37529.1 | - | 3 | 291 | |||
X54469.1 | - | 3 | 227 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
