siRNA Details
Gene Symbol: TAOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016151.3 | NP_057235.2 | 16 | 3224 | ||
XM_011545982.1 | XP_011544284.1 | 16 | 3471 | |||
XM_011545983.2 | XP_011544285.1 | 16 | 3468 | |||
GenBank mRNA | AF061943.1 | - | 16 | 3165 | ||
AL137701.1 | - | 8 | 3061 | |||
BC051798.1 | - | 16 | 2981 | |||
BC136653.1 | - | 16 | 2909 | 9020643 | ||
BC136655.1 | - | 16 | 2909 | 9020645 | ||
BC142663.1 | - | 16 | 2564 | 40148112 | ||
BC151221.1 | - | 16 | 2564 | 40148752 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
