siRNA Details
Gene Symbol: TAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003182.2 | NP_003173.1 | 7 | 982 | ||
NM_013996.2 | NP_054702.1 | 6 | 928 | |||
NM_013997.2 | NP_054703.1 | 6 | 937 | |||
NM_013998.2 | NP_054704.1 | 5 | 883 | |||
GenBank mRNA | AK222957.1 | - | 7 | 857 | ||
BC018047.1 | - | 7 | 856 | 4792746 | ||
U37529.1 | - | 7 | 881 | |||
X54469.1 | - | 7 | 817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
