siRNA Details
Gene Symbol: SEC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136026.2 | NP_001129498.1 | 3 | 332 | ||
NM_001136232.2 | NP_001129704.1 | 2 | 186 | |||
NM_001278946.1 | NP_001265875.1 | 2 | 154 | |||
NM_030673.3 | NP_109598.2 | 3 | 188 | |||
NM_183352.2 | NP_899195.1 | 2 | 154 | |||
XM_005265379.2 | XP_005265436.1 | 3 | 205 | |||
XM_017007019.1 | XP_016862508.1 | 3 | 242 | |||
XM_017007020.1 | XP_016862509.1 | 2 | 49 | |||
XM_017007021.1 | XP_016862510.1 | 2 | 59 | |||
GenBank mRNA | AF052155.1 | - | 2 | 186 | ||
AK223019.1 | - | 2 | 39 | |||
AK294662.1 | - | 2 | 59 | |||
AK301999.1 | - | 3 | 256 | |||
BC002634.2 | - | 3 | 69 | 3605545 | ||
BC091506.1 | - | 2 | 64 | 30408210 | ||
BE621430.1 | - | 2 | 140 | |||
CN344033.1 | - | 2 | 166 | |||
EF036494.1 | - | 2 | 53 | |||
L09260.1 | - | 2 | 31 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
