siRNA Details
Gene Symbol: NECAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031231.3 | NP_112508.3 | 12, 13 | 1174 | ||
NM_031232.3 | NP_112509.3 | 11, 12 | 1276 | |||
XM_005260510.1 | XP_005260567.1 | 11, 12 | 1090 | |||
XM_011528991.1 | XP_011527293.1 | 8, 9 | 800 | |||
XM_017028015.1 | XP_016883504.1 | 8, 9 | 799 | |||
XM_017028016.1 | XP_016883505.1 | 8, 9 | 728 | |||
GenBank mRNA | AB039947.1 | - | 9, 10 | 978 | ||
AF193759.1 | - | 12, 13 | 1081 | |||
AF409141.1 | - | 12, 13 | 1122 | |||
AK023706.1 | - | 11, 12 | 1380 | |||
AK291895.1 | - | 12, 13 | 1104 | |||
BC047673.1 | - | 12, 13 | 1129 | 5755231 | ||
CD580591.2 | - | 1, 2 | 114 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
