siRNA Details
Gene Symbol: NECAB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031231.3 | NP_112508.3 | 3 | 278 | ||
NM_031232.3 | NP_112509.3 | 3 | 278 | |||
XM_005260510.1 | XP_005260567.1 | 3 | 248 | |||
XM_011528992.2 | XP_011527294.1 | 3 | 248 | |||
GenBank mRNA | AF193759.1 | - | 3 | 185 | ||
AF409141.1 | - | 3 | 226 | |||
AK291895.1 | - | 3 | 208 | |||
AK309285.1 | - | 3 | 240 | |||
AL519842.3 | - | 3 | 278 | |||
BC047673.1 | - | 3 | 233 | 5755231 | ||
BQ431858.1 | - | 3 | 224 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
