siRNA Details
Gene Symbol: SULT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001055.3 | NP_001046.2 | 8 | 1075 | ||
NM_177529.2 | NP_803565.1 | 8 | 1106 | |||
NM_177530.2 | NP_803566.1 | 8 | 1231 | |||
NM_177534.2 | NP_803878.1 | 7 | 1430 | |||
NM_177536.3 | NP_803880.1 | 6 | 1112 | |||
XM_017023604.1 | XP_016879093.1 | 9 | 1178 | |||
XM_017023605.1 | XP_016879094.1 | 9 | 1175 | |||
XM_017023606.1 | XP_016879095.1 | 9 | 1160 | |||
XM_017023607.1 | XP_016879096.1 | 11 | 1721 | |||
XM_017023608.1 | XP_016879097.1 | 8 | 1093 | |||
XM_017023609.1 | XP_016879098.1 | 8 | 1143 | |||
XM_017023610.1 | XP_016879099.1 | 8 | 1174 | |||
XM_017023611.1 | XP_016879100.1 | 10 | 1648 | |||
XM_017023612.1 | XP_016879101.1 | 11 | 1572 | |||
XM_017023613.1 | XP_016879102.1 | 9 | 1387 | |||
GenBank mRNA | AB062428.1 | - | 7 | 1013 | ||
AB209149.1 | - | 7790 | ||||
BC000923.2 | - | 8 | 1025 | 3447900 | ||
BC110887.1 | - | 11 | 1488 | 5190558 | ||
L10819.1 | - | 8 | 995 | |||
L19955.1 | - | 7 | 998 | |||
U09031.1 | - | 7 | 998 | |||
U26309.1 | - | 7 | 1021 | |||
X78283.1 | - | 8 | 1116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
