siRNA Details
Gene Symbol: SULT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001055.3 | NP_001046.2 | 8 | 1199 | ||
NM_177529.2 | NP_803565.1 | 8 | 1230 | |||
NM_177530.2 | NP_803566.1 | 8 | 1355 | |||
NM_177534.2 | NP_803878.1 | 7 | 1554 | |||
NM_177536.3 | NP_803880.1 | 6 | 1236 | |||
XM_017023604.1 | XP_016879093.1 | 9 | 1302 | |||
XM_017023605.1 | XP_016879094.1 | 9 | 1299 | |||
XM_017023606.1 | XP_016879095.1 | 9 | 1284 | |||
XM_017023607.1 | XP_016879096.1 | 11 | 1845 | |||
XM_017023608.1 | XP_016879097.1 | 8 | 1217 | |||
XM_017023609.1 | XP_016879098.1 | 8 | 1267 | |||
XM_017023610.1 | XP_016879099.1 | 8 | 1298 | |||
XM_017023611.1 | XP_016879100.1 | 10 | 1772 | |||
XM_017023612.1 | XP_016879101.1 | 11 | 1696 | |||
XM_017023613.1 | XP_016879102.1 | 9 | 1511 | |||
GenBank mRNA | AB062428.1 | - | 7 | 1137 | ||
AB209149.1 | - | 7914 | ||||
BC000923.2 | - | 8 | 1149 | 3447900 | ||
BC110887.1 | - | 11 | 1612 | 5190558 | ||
L10819.1 | - | 8 | 1119 | |||
L19955.1 | - | 7 | 1122 | |||
L19999.1 | - | 8 | 1150 | |||
U09031.1 | - | 8 | 1122 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
