siRNA Details
Gene Symbol: SULT1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177536.3 | NP_803880.1 | 2 | 485 | ||
XM_017023607.1 | XP_016879096.1 | 2 | 384 | |||
XM_017023611.1 | XP_016879100.1 | 2 | 384 | |||
XM_017023612.1 | XP_016879101.1 | 2 | 384 | |||
XM_017023613.1 | XP_016879102.1 | 2 | 384 | |||
GenBank mRNA | AL558375.3 | - | 2 | 483 | ||
BC110887.1 | - | 2 | 151 | 5190558 | ||
BI460081.1 | - | 2 | 448 |
Gene Symbol: MPV17L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128423.1 | NP_001121895.1 | 2 | 483 | ||
GenBank mRNA | AK293380.1 | - | 2 | 458 | ||
DQ004255.1 | - | 2 | 483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
