siRNA Details
Gene Symbol: ZFP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320666.1 | NP_001307595.1 | 6 | 2577 | ||
NM_001320667.1 | NP_001307596.1 | 6 | 2483 | |||
NM_001320668.1 | NP_001307597.1 | 6 | 2353 | |||
NM_001320669.1 | NP_001307598.1 | 6 | 2291 | |||
NM_001320670.1 | NP_001307599.1 | 6 | 2288 | |||
NM_001320671.1 | NP_001307600.1 | 6 | 2194 | |||
NM_014898.2 | NP_055713.1 | 6 | 2524 | |||
GenBank mRNA | AB023178.1 | - | 6 | 2524 | ||
BC041087.1 | - | 6 | 2553 | 5579247 | ||
BC050542.1 | - | 6 | 2482 | |||
GU388430.1 | - | 6 | 2353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
