siRNA Details
Gene Symbol: GIPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005716.3 | NP_005707.1 | 9 | 1915 | ||
NM_202468.2 | NP_974197.1 | 8 | 1859 | |||
NM_202469.2 | NP_974198.1 | 7 | 1541 | |||
NM_202470.2 | NP_974199.1 | 7 | 1771 | |||
NM_202494.2 | NP_974223.1 | 6 | 1453 | |||
XM_017026147.1 | XP_016881636.1 | 8 | 1806 | |||
XM_017026148.1 | XP_016881637.1 | 8 | 1570 | |||
GenBank mRNA | AF028824.1 | - | 6 | 1399 | ||
AF089816.1 | - | 7 | 1719 | |||
AK022585.1 | - | 6 | 1630 | |||
AK093820.1 | - | 3 | 2045 | |||
BC000410.2 | - | 9 | 1887 | 2960992 | ||
BC004226.2 | - | 6 | 1530 | |||
BC012810.2 | - | 9 | 1887 | 2960992 | ||
BC016169.1 | - | 9 | 1887 | 3530664 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
