siRNA Details
Gene Symbol: LAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014146.3 | NP_054865.2 | 10 | 770 | ||
NM_032463.2 | NP_115852.1 | 11 | 889 | |||
NM_032464.2 | NP_115853.2 | 11 | 959 | |||
XM_011516558.2 | XP_011514860.1 | 11 | 799 | |||
GenBank mRNA | AF161531.1 | - | 10 | 519 | ||
AF252611.1 | - | 11 | 710 | |||
AF252612.1 | - | 9 | 465 | |||
AF252613.1 | - | 10 | 506 | |||
AF252614.1 | - | 11 | 640 | |||
AF257135.1 | - | 9 | 441 | |||
AK002099.1 | - | 11 | 749 | |||
AK092904.1 | - | 11 | 1100 | |||
AK225150.1 | - | 10 | 521 | |||
AK290074.1 | - | 11 | 637 | |||
AK290916.1 | - | 10 | 518 | |||
AY190023.1 | - | 9 | 451 | |||
BC001609.1 | - | 11 | 771 | 3543310 | ||
BC009204.2 | - | 9 | 443 | 4054137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
