siRNA Details
Gene Symbol: LAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014146.3 | NP_054865.2 | 13 | 1710 | ||
NM_032463.2 | NP_115852.1 | 14 | 1829 | |||
NM_032464.2 | NP_115853.2 | 14 | 1899 | |||
XM_011516558.2 | XP_011514860.1 | 14 | 1739 | |||
GenBank mRNA | AF086239.1 | - | 1 | 166 | ||
AF252611.1 | - | 14 | 1650 | |||
AF252612.1 | - | 12 | 1405 | |||
AF252613.1 | - | 13 | 1446 | |||
AF252614.1 | - | 14 | 1580 | |||
AF257135.1 | - | 12 | 1381 | |||
AK002099.1 | - | 14 | 1689 | |||
AK092904.1 | - | 14 | 2040 | |||
AK225150.1 | - | 12 | 1146 | |||
BC001609.1 | - | 14 | 1711 | 3543310 | ||
BC009204.2 | - | 12 | 1383 | 4054137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
