siRNA Details
Gene Symbol: IKZF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257408.1 | NP_001244337.1 | 7 | 2015 | ||
NM_001257409.1 | NP_001244338.1 | 6 | 1898 | |||
NM_001257410.1 | NP_001244339.1 | 7 | 1856 | |||
NM_001257411.1 | NP_001244340.1 | 6 | 1688 | |||
NM_001257412.1 | NP_001244341.1 | 5 | 1571 | |||
NM_001257413.1 | NP_001244342.1 | 4 | 1454 | |||
NM_001257414.1 | NP_001244343.1 | 5 | 1715 | |||
NM_001284514.1 | NP_001271443.1 | 9 | 2243 | |||
NM_001284515.1 | NP_001271444.1 | 8 | 2075 | |||
NM_001284516.1 | NP_001271445.1 | 3 | 1687 | |||
NM_012481.4 | NP_036613.2 | 8 | 2117 | |||
NM_183228.2 | NP_899051.1 | 7 | 1949 | |||
NM_183229.2 | NP_899052.1 | 7 | 2000 | |||
NM_183230.2 | NP_899053.1 | 7 | 2000 | |||
NM_183231.2 | NP_899054.1 | 6 | 1832 | |||
NM_183232.2 | NP_899055.1 | 6 | 1883 | |||
GenBank mRNA | BC032707.1 | - | 8 | 2106 | 5497068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
