siRNA Details
Gene Symbol: IKZF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257410.1 | NP_001244339.1 | 3 | 135 | ||
NM_001257411.1 | NP_001244340.1 | 3 | 135 | |||
NM_001257412.1 | NP_001244341.1 | 3 | 135 | |||
NM_001257413.1 | NP_001244342.1 | 3 | 135 | |||
NM_001257414.1 | NP_001244343.1 | 3 | 135 | |||
NM_001284514.1 | NP_001271443.1 | 3 | 135 | |||
NM_001284515.1 | NP_001271444.1 | 3 | 135 | |||
NM_012481.4 | NP_036613.2 | 3 | 135 | |||
NM_183228.2 | NP_899051.1 | 3 | 135 | |||
NM_183229.2 | NP_899052.1 | 3 | 135 | |||
NM_183230.2 | NP_899053.1 | 3 | 135 | |||
NM_183231.2 | NP_899054.1 | 3 | 135 | |||
NM_183232.2 | NP_899055.1 | 3 | 135 | |||
GenBank mRNA | AF129512.1 | - | 3 | 135 | ||
AJ292565.1 | - | 2 | 73 | |||
AJ292566.1 | - | 2 | 73 | |||
AJ292567.1 | - | 2 | 73 | |||
AJ292568.1 | - | 2 | 73 | |||
AJ292569.1 | - | 2 | 73 | |||
AY377976.1 | - | 3 | 135 | |||
AY377977.1 | - | 3 | 135 | |||
AY377978.1 | - | 3 | 135 | |||
AY377979.1 | - | 3 | 135 | |||
AY377980.1 | - | 3 | 135 | |||
AY377981.1 | - | 3 | 135 | |||
AY377982.1 | - | 3 | 135 | |||
BC032707.1 | - | 3 | 124 | 5497068 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
