siRNA Details
Gene Symbol: ELMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318253.1 | NP_001305182.1 | 13, 14 | 1769 | ||
NM_133171.4 | NP_573403.1 | 13, 14 | 1701 | |||
NM_182764.2 | NP_877496.1 | 12, 13 | 1597 | |||
XM_005260496.3 | XP_005260553.1 | 13, 14 | 1266 | |||
XM_005260498.3 | XP_005260555.1 | 12, 13 | 1628 | |||
XM_005260499.3 | XP_005260556.1 | 12, 13 | 1599 | |||
XM_005260500.3 | XP_005260557.1 | 11, 12 | 1127 | |||
XM_005260501.4 | XP_005260558.1 | 12, 13 | 1137 | |||
XM_006723854.3 | XP_006723917.1 | 12, 13 | 1200 | |||
XM_017028009.1 | XP_016883498.1 | 12, 13 | 1572 | |||
XM_017028010.1 | XP_016883499.1 | 11, 12 | 1075 | |||
XM_017028011.1 | XP_016883500.1 | 11, 12 | 1528 | |||
GenBank mRNA | AB058737.1 | - | 12, 13 | 1212 | ||
AF398886.1 | - | 11, 12 | 1140 | |||
AF417861.1 | - | 11, 12 | 1072 | |||
AK021718.1 | - | 2, 3 | 268 | |||
AK023103.1 | - | 1, 2 | 28 | |||
AK293861.1 | - | 11, 12 | 1165 | |||
AK299318.1 | - | 15, 16 | 1312 | |||
AK302710.1 | - | 3, 4 | 504 | |||
AL713765.1 | - | 2, 3 | 246 | |||
BC000143.4 | - | 12, 13 | 1277 | 3047992 | ||
BC021079.2 | - | 2, 3 | 264 | |||
BX648819.1 | - | 12, 13 | 1178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
