siRNA Details
Gene Symbol: ELMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318253.1 | NP_001305182.1 | 15, 16 | 1987 | ||
NM_133171.4 | NP_573403.1 | 15, 16 | 1919 | |||
NM_182764.2 | NP_877496.1 | 14, 15 | 1815 | |||
XM_005260496.3 | XP_005260553.1 | 15, 16 | 1484 | |||
XM_005260498.3 | XP_005260555.1 | 14, 15 | 1846 | |||
XM_005260499.3 | XP_005260556.1 | 14, 15 | 1817 | |||
XM_005260500.3 | XP_005260557.1 | 13, 14 | 1345 | |||
XM_005260501.4 | XP_005260558.1 | 14, 15 | 1355 | |||
XM_006723854.3 | XP_006723917.1 | 14, 15 | 1418 | |||
XM_017028009.1 | XP_016883498.1 | 14, 15 | 1790 | |||
XM_017028010.1 | XP_016883499.1 | 13, 14 | 1293 | |||
XM_017028011.1 | XP_016883500.1 | 13, 14 | 1746 | |||
GenBank mRNA | AB058737.1 | - | 14, 15 | 1430 | ||
AF398886.1 | - | 13, 14 | 1358 | |||
AF417861.1 | - | 13, 14 | 1290 | |||
AK021718.1 | - | 4, 5 | 486 | |||
AK022731.1 | - | 2, 3 | 201 | |||
AK023103.1 | - | 3, 4 | 246 | |||
AK293861.1 | - | 13, 14 | 1383 | |||
AK299318.1 | - | 17, 18 | 1530 | |||
AK302710.1 | - | 5, 6 | 722 | |||
AL713765.1 | - | 4, 5 | 464 | |||
BC000143.4 | - | 14, 15 | 1495 | 3047992 | ||
BC021079.2 | - | 4, 5 | 482 | |||
BX648819.1 | - | 14, 15 | 1396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
