siRNA Details
Gene Symbol: ELMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318253.1 | NP_001305182.1 | 13 | 1670 | ||
NM_133171.4 | NP_573403.1 | 13 | 1602 | |||
NM_182764.2 | NP_877496.1 | 12 | 1498 | |||
XM_005260496.3 | XP_005260553.1 | 13 | 1167 | |||
XM_005260498.3 | XP_005260555.1 | 12 | 1529 | |||
XM_005260499.3 | XP_005260556.1 | 12 | 1500 | |||
XM_005260500.3 | XP_005260557.1 | 11 | 1028 | |||
XM_005260501.4 | XP_005260558.1 | 12 | 1038 | |||
XM_006723854.3 | XP_006723917.1 | 12 | 1101 | |||
XM_017028009.1 | XP_016883498.1 | 12 | 1473 | |||
XM_017028010.1 | XP_016883499.1 | 11 | 976 | |||
XM_017028011.1 | XP_016883500.1 | 11 | 1429 | |||
GenBank mRNA | AB058737.1 | - | 12 | 1113 | ||
AF398886.1 | - | 11 | 1041 | |||
AF417861.1 | - | 11 | 973 | |||
AK021718.1 | - | 2 | 169 | |||
AK057032.1 | - | 13 | 1126 | |||
AK293861.1 | - | 11 | 1066 | |||
AK299318.1 | - | 15 | 1213 | |||
AK302710.1 | - | 3 | 405 | |||
AL713765.1 | - | 2 | 147 | |||
BC000143.4 | - | 12 | 1178 | 3047992 | ||
BC021079.2 | - | 2 | 165 | |||
BX648819.1 | - | 12 | 1079 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
