siRNA Details
Gene Symbol: RFC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278791.1 | NP_001265720.1 | 4 | 497 | ||
NM_001278793.1 | NP_001265722.1 | 4 | 497 | |||
NM_002914.4 | NP_002905.2 | 4 | 395 | |||
NM_181471.2 | NP_852136.1 | 4 | 395 | |||
XM_017012492.1 | XP_016867981.1 | 4 | 373 | |||
GenBank mRNA | AB451243.1 | - | 4 | 320 | ||
AB451368.1 | - | 4 | 320 | |||
BC002813.1 | - | 4 | 325 | 3637774 | ||
BM563468.1 | - | 4 | 426 | |||
BQ070526.1 | - | 4 | 345 | |||
BQ674131.1 | - | 4 | 348 | |||
BT007058.1 | - | 4 | 320 | |||
HY046046.1 | - | 4 | 396 | |||
HY056452.1 | - | 4 | 389 | |||
M87338.1 | - | 527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
