siRNA Details
Gene Symbol: CSMD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052900.2 | NP_443132.3 | 22 | 3621 | ||
NM_198123.1 | NP_937756.1 | 23 | 3933 | |||
NM_198124.1 | NP_937757.1 | 24 | 3825 | |||
XM_011516816.2 | XP_011515118.1 | 22 | 4107 | |||
XM_017013008.1 | XP_016868497.1 | 22 | 4341 | |||
XM_017013009.1 | XP_016868498.1 | 23 | 4419 | |||
XM_017013010.1 | XP_016868499.1 | 21 | 4029 | |||
XM_017013011.1 | XP_016868500.1 | 21 | 4029 | |||
XM_017013012.1 | XP_016868501.1 | 21 | 3392 | |||
GenBank mRNA | AB067481.2 | - | 10 | 1794 | ||
AB114604.1 | - | 23 | 3933 | |||
AB114605.1 | - | 24 | 3825 | |||
AY210419.1 | - | 22 | 3492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
