siRNA Details
Gene Symbol: PTTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282382.1 | NP_001269311.1 | 5 | 1010 | ||
NM_001282383.1 | NP_001269312.1 | 6 | 864 | |||
NM_004219.3 | NP_004210.1 | 6 | 703 | |||
GenBank mRNA | AF075242.1 | - | 5 | 627 | ||
AF095287.1 | - | 6 | 643 | |||
AJ223953.1 | - | 5 | 712 | |||
BC101834.1 | - | 6 | 636 | 8069340 | ||
BC112071.1 | - | 6 | 636 | 8327539 | ||
BU182149.1 | - | 6 | 860 |
Gene Symbol: PTTG3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002734.2 | 1 | 636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
