siRNA Details
Gene Symbol: KBTBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152433.3 | NP_689646.2 | 3 | 1308 | ||
NM_198439.2 | NP_940841.1 | 4 | 1509 | |||
XM_006718767.3 | XP_006718830.1 | 4 | 1539 | |||
XM_011542618.2 | XP_011540920.1 | 3 | 1153 | |||
XM_011542619.2 | XP_011540921.1 | 3 | 1291 | |||
GenBank mRNA | AK092993.1 | - | 3 | 911 | ||
AK289915.1 | - | 4 | 1119 | |||
BC117466.1 | - | 3 | 1007 | 40126017 | ||
BX640672.1 | - | 3 | 1029 | |||
BX647830.1 | - | 2 | 674 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
