siRNA Details
Gene Symbol: FBXO25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012173.3 | NP_036305.2 | 6 | 713 | ||
NM_183420.1 | NP_904356.1 | 7 | 763 | |||
NM_183421.1 | NP_904357.1 | 7 | 763 | |||
XM_011534748.2 | XP_011533050.1 | 7 | 536 | |||
XM_011534749.1 | XP_011533051.1 | 6 | 473 | |||
XM_017013313.1 | XP_016868802.1 | 7 | 587 | |||
XM_017013314.1 | XP_016868803.1 | 6 | 665 | |||
GenBank mRNA | AB070270.1 | - | 6 | 497 | ||
AF174605.1 | - | 3 | 239 | |||
AF419858.1 | - | 7 | 763 | |||
AK302334.1 | - | 7 | 522 | |||
BC020249.2 | - | 5 | 455 | 4652811 | ||
BC050393.1 | - | 7 | 616 | 5769518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
