siRNA Details
Gene Symbol: FBXO25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012173.3 | NP_036305.2 | 9 | 1617 | ||
NM_183420.1 | NP_904356.1 | 10 | 1667 | |||
NM_183421.1 | NP_904357.1 | 11 | 1694 | |||
XM_011534748.2 | XP_011533050.1 | 11 | 1467 | |||
XM_017013313.1 | XP_016868802.1 | 11 | 1518 | |||
GenBank mRNA | AA316216.1 | - | 1 | 242 | ||
BC050393.1 | - | 11 | 1547 | 5769518 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
