siRNA Details
Gene Symbol: PKIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006823.3 | NP_006814.1 | 4 | 2098 | ||
NM_181839.2 | NP_862822.1 | 3 | 1969 | |||
XM_017013620.1 | XP_016869109.1 | 5 | 1973 | |||
XM_017013621.1 | XP_016869110.1 | 4 | 1939 | |||
XM_017013622.1 | XP_016869111.1 | 3 | 1704 | |||
GenBank mRNA | BC022265.2 | - | 2 | 1600 | 4288676 | |
S76965.1 | - | 5 | 2073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
