siRNA Details
Gene Symbol: RPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000971.3 | NP_000962.2 | 1 | 789 | ||
XM_006716463.2 | XP_006716526.1 | 7 | 1382 | |||
GenBank mRNA | AK225246.1 | - | 1 | 788 | ||
AK225254.1 | - | 1 | 787 | |||
AK291119.1 | - | 7 | 787 | |||
BC006095.1 | - | 1 | 777 | 2821364 | ||
BC008850.2 | - | 1 | 814 | 4138113 | ||
BC009599.1 | - | 1 | 789 | 4134557 | ||
BC071671.1 | - | 1 | 770 | 4392472 | ||
BC071894.1 | - | 1 | 774 | 2906310 | ||
BC071895.1 | - | 1 | 772 | 6568654 | ||
BC087837.1 | - | 1 | 773 | 6170371 | ||
L16558.1 | - | 1 | 767 | |||
X52967.1 | - | 7 | 788 | |||
X57958.1 | - | 1 | 804 | |||
X57959.1 | - | 1 | 777 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
