siRNA Details
Gene Symbol: RPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000971.3 | NP_000962.2 | 1 | 816 | ||
XM_006716463.2 | XP_006716526.1 | 7 | 1409 | |||
GenBank mRNA | AK225246.1 | - | 1 | 815 | ||
AK225254.1 | - | 1 | 814 | |||
AK291119.1 | - | 7 | 814 | |||
BC008850.2 | - | 1 | 841 | 4138113 | ||
BC009599.1 | - | 1 | 816 | 4134557 | ||
BC071671.1 | - | 1 | 797 | 4392472 | ||
BC071894.1 | - | 1 | 801 | 2906310 | ||
BC071895.1 | - | 1 | 799 | 6568654 | ||
BC087837.1 | - | 1 | 800 | 6170371 | ||
L16558.1 | - | 1 | 794 | |||
X52967.1 | - | 7 | 815 | |||
X57958.1 | - | 1 | 831 | |||
X57959.1 | - | 1 | 804 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
