siRNA Details
Gene Symbol: AMPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001635.3 | NP_001626.1 | 18, 19 | 1835 | ||
NM_139316.2 | NP_647477.1 | 17, 18 | 1709 | |||
XM_006715689.3 | XP_006715752.1 | 17, 18 | 3372 | |||
XM_006715690.3 | XP_006715753.1 | 17, 18 | 2154 | |||
XM_011515271.2 | XP_011513573.1 | 18, 19 | 2280 | |||
XM_017011995.1 | XP_016867484.1 | 18, 19 | 3498 | |||
XM_017011996.1 | XP_016867485.1 | 17, 18 | 2460 | |||
GenBank mRNA | AF034996.1 | - | 17, 18 | 1562 | ||
AF498092.1 | - | 6, 7 | 890 | |||
AF498093.1 | - | 4, 5 | 847 | |||
AF498094.1 | - | 4, 5 | 1759 | |||
AF498095.1 | - | 5, 6 | 646 | |||
AF498096.1 | - | 9, 10 | 1285 | |||
AK291447.1 | - | 18, 19 | 1639 | |||
BC034376.1 | - | 18, 19 | 1626 | 5181100 | ||
U07616.1 | - | 18, 19 | 1729 | |||
X81438.1 | - | 18, 19 | 1693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
