siRNA Details
Gene Symbol: AMPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001635.3 | NP_001626.1 | 18 | 1689 | ||
NM_139316.2 | NP_647477.1 | 17 | 1563 | |||
XM_006715689.3 | XP_006715752.1 | 17 | 3226 | |||
XM_006715690.3 | XP_006715753.1 | 17 | 2008 | |||
XM_011515271.2 | XP_011513573.1 | 18 | 2134 | |||
XM_017011995.1 | XP_016867484.1 | 18 | 3352 | |||
XM_017011996.1 | XP_016867485.1 | 17 | 2314 | |||
GenBank mRNA | AF034996.1 | - | 17 | 1416 | ||
AF498092.1 | - | 6 | 744 | |||
AF498093.1 | - | 4 | 701 | |||
AF498094.1 | - | 4 | 1613 | |||
AF498095.1 | - | 5 | 500 | |||
AF498096.1 | - | 9 | 1139 | |||
AK291447.1 | - | 18 | 1493 | |||
BC034376.1 | - | 18 | 1480 | 5181100 | ||
U07616.1 | - | 18 | 1583 | |||
X81438.1 | - | 18 | 1547 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
