siRNA Details
Gene Symbol: AMPH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001635.3 | NP_001626.1 | 18 | 1737 | ||
NM_139316.2 | NP_647477.1 | 17 | 1611 | |||
XM_006715689.3 | XP_006715752.1 | 17 | 3274 | |||
XM_006715690.3 | XP_006715753.1 | 17 | 2056 | |||
XM_011515271.2 | XP_011513573.1 | 18 | 2182 | |||
XM_017011995.1 | XP_016867484.1 | 18 | 3400 | |||
XM_017011996.1 | XP_016867485.1 | 17 | 2362 | |||
GenBank mRNA | AF034996.1 | - | 17 | 1464 | ||
AF498092.1 | - | 6 | 792 | |||
AF498093.1 | - | 4 | 749 | |||
AF498094.1 | - | 4 | 1661 | |||
AF498095.1 | - | 5 | 548 | |||
AF498096.1 | - | 9 | 1187 | |||
AK291447.1 | - | 18 | 1541 | |||
BC034376.1 | - | 18 | 1528 | 5181100 | ||
U07616.1 | - | 18 | 1631 | |||
X81438.1 | - | 18 | 1595 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
