siRNA Details
Gene Symbol: CCDC50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174908.3 | NP_777568.1 | 11 | 1693 | ||
NM_178335.2 | NP_848018.1 | 12 | 2221 | |||
GenBank mRNA | AJ416916.2 | - | 11 | 1693 | ||
AJ557013.1 | - | 12 | 2221 | |||
BC065004.1 | - | 11 | 1190 | 6049272 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
