siRNA Details
Gene Symbol: CD59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000611.5 | NP_000602.1 | 4 | 1644 | ||
NM_001127223.1 | NP_001120695.1 | 3 | 1810 | |||
NM_001127225.1 | NP_001120697.1 | 4 | 1703 | |||
NM_001127226.1 | NP_001120698.1 | 4 | 1638 | |||
NM_001127227.1 | NP_001120699.1 | 4 | 1582 | |||
NM_203329.2 | NP_976074.1 | 5 | 1760 | |||
NM_203330.2 | NP_976075.1 | 6 | 1805 | |||
NM_203331.2 | NP_976076.1 | 5 | 1689 | |||
GenBank mRNA | AK128571.1 | - | 2 | 3601 | ||
BC001506.2 | - | 4 | 1568 | 2988140 | ||
M34671.1 | - | 3 | 1556 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
