siRNA Details
Gene Symbol: CD59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000611.5 | NP_000602.1 | 4 | 5372 | ||
NM_001127223.1 | NP_001120695.1 | 3 | 5538 | |||
NM_001127225.1 | NP_001120697.1 | 4 | 5431 | |||
NM_001127226.1 | NP_001120698.1 | 4 | 5366 | |||
NM_001127227.1 | NP_001120699.1 | 4 | 5310 | |||
NM_203329.2 | NP_976074.1 | 5 | 5488 | |||
NM_203330.2 | NP_976075.1 | 6 | 5533 | |||
NM_203331.2 | NP_976076.1 | 5 | 5417 | |||
GenBank mRNA | BM911556.1 | - | 1 | 323 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
