siRNA Details
Gene Symbol: RNF38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022781.4 | NP_073618.3 | 12 | 2079 | ||
NM_194328.2 | NP_919309.1 | 12 | 2246 | |||
NM_194329.2 | NP_919310.1 | 11 | 1929 | |||
NM_194330.2 | NP_919311.1 | 12 | 1977 | |||
NM_194332.2 | NP_919313.1 | 11 | 2096 | |||
XM_005251366.3 | XP_005251423.1 | 12 | 2083 | |||
XM_005251367.3 | XP_005251424.1 | 12 | 1981 | |||
XM_005251368.3 | XP_005251425.1 | 12 | 2095 | |||
XM_006716721.3 | XP_006716784.1 | 11 | 1943 | |||
XM_011517712.2 | XP_011516014.1 | 13 | 2019 | |||
XM_011517713.2 | XP_011516015.1 | 12 | 2083 | |||
XM_017014294.1 | XP_016869783.1 | 11 | 2000 | |||
XM_017014295.1 | XP_016869784.1 | 12 | 2110 | |||
XM_017014296.1 | XP_016869785.1 | 11 | 1680 | |||
XM_017014297.1 | XP_016869786.1 | 11 | 1676 | |||
GenBank mRNA | AF394047.1 | - | 12 | 2201 | ||
AK024996.1 | - | 10 | 1589 | |||
AK025466.1 | - | 1 | 104 | |||
AL120640.1 | - | 1 | 195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
