siRNA Details
Gene Symbol: FCAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002000.3 | NP_001991.1 | 5 | 1497 | ||
NM_133269.3 | NP_579803.1 | 5 | 1431 | |||
NM_133271.3 | NP_579805.1 | 4 | 1209 | |||
NM_133272.3 | NP_579806.1 | 4 | 1461 | |||
NM_133273.3 | NP_579807.1 | 3 | 1173 | |||
NM_133274.3 | NP_579808.1 | 4 | 1387 | |||
NM_133277.3 | NP_579811.1 | 3 | 1170 | |||
NM_133278.3 | NP_579812.1 | 4 | 1395 | |||
XM_011526625.2 | XP_011524927.1 | 5 | 1421 | |||
XM_017026473.1 | XP_016881962.1 | 5 | 1470 | |||
XM_017026474.1 | XP_016881963.1 | 4 | 1253 | |||
GenBank mRNA | BC027953.1 | - | 5 | 1326 | 5211453 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
