siRNA Details
Gene Symbol: RBAK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204456.1 | NP_001191385.1 | 6 | 635 | ||
NM_021163.3 | NP_066986.1 | 5 | 830 | |||
GenBank mRNA | AK293069.1 | - | 6 | 635 | ||
BC136675.1 | - | 5 | 483 | 9020665 | ||
BC136676.1 | - | 5 | 483 | 9020666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
