siRNA Details
Gene Symbol: FLRT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013281.3 | NP_037413.1 | 2 | 3625 | ||
NM_198391.2 | NP_938205.1 | 3 | 3819 | |||
XM_005260682.4 | XP_005260739.1 | 3 | 4337 | |||
XM_011529204.2 | XP_011527506.1 | 4 | 4474 | |||
XM_011529205.2 | XP_011527507.1 | 4 | 4136 | |||
GenBank mRNA | AB040902.1 | - | 2 | 3513 | ||
AK027670.1 | - | 1 | 2732 | |||
AK074909.1 | - | 2 | 3492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
