siRNA Details
Gene Symbol: ATRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000489.4 | NP_000480.3 | 35 | 9571 | ||
NM_138270.3 | NP_612114.2 | 34 | 9457 | |||
XM_005262153.4 | XP_005262210.2 | 35 | 9568 | |||
XM_005262154.4 | XP_005262211.2 | 36 | 9484 | |||
XM_005262155.4 | XP_005262212.2 | 35 | 9672 | |||
XM_005262156.4 | XP_005262213.2 | 34 | 9424 | |||
XM_005262157.4 | XP_005262214.2 | 35 | 9367 | |||
XM_006724666.3 | XP_006724729.1 | 34 | 9454 | |||
XM_006724667.3 | XP_006724730.1 | 33 | 9310 | |||
XM_017029601.1 | XP_016885090.1 | 36 | 9481 | |||
XM_017029602.1 | XP_016885091.1 | 35 | 9652 | |||
XM_017029603.1 | XP_016885092.1 | 34 | 9421 | |||
XM_017029604.1 | XP_016885093.1 | 35 | 9370 | |||
XM_017029605.1 | XP_016885094.1 | 36 | 9585 | |||
XM_017029606.1 | XP_016885095.1 | 34 | 9558 | |||
XM_017029607.1 | XP_016885096.1 | 34 | 9555 | |||
XM_017029608.1 | XP_016885097.1 | 33 | 9307 | |||
XM_017029609.1 | XP_016885098.1 | 35 | 9471 | |||
XM_017029610.1 | XP_016885099.1 | 35 | 9468 | |||
XM_017029611.1 | XP_016885100.1 | 34 | 9223 | |||
GenBank mRNA | AA484729.1 | - | 1 | 248 | ||
U72936.2 | - | 36 | 9642 | |||
U72937.2 | - | 35 | 9520 | |||
U72938.2 | - | 34 | 9406 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
