siRNA Details
Gene Symbol: PLEKHG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042663.1 | NP_001036128.1 | 22 | 3686 | ||
NM_001042664.1 | NP_001036129.1 | 21 | 3640 | |||
NM_001042665.1 | NP_001036130.1 | 21 | 3623 | |||
NM_001265592.1 | NP_001252521.1 | 22 | 3719 | |||
NM_001265593.1 | NP_001252522.1 | 21 | 3650 | |||
NM_001265594.1 | NP_001252523.1 | 22 | 3454 | |||
NM_020631.4 | NP_065682.2 | 21 | 3569 | |||
NM_198681.3 | NP_941374.2 | 22 | 4198 | |||
GenBank mRNA | AB018263.2 | - | 22 | 3674 | ||
AB097001.1 | - | 20 | 3409 | |||
AK024676.1 | - | 21 | 3568 | |||
AK091201.1 | - | 9 | 2619 | |||
AK096347.1 | - | 22 | 4146 | |||
AK125550.1 | - | 22 | 3652 | |||
AK128021.1 | - | 1 | 1914 | |||
AK131074.1 | - | 19 | 3893 | |||
AK299523.1 | - | 21 | 3650 | |||
BC015231.1 | - | 22 | 3430 | 3900065 | ||
BC036671.2 | - | 22 | 3506 | |||
BF033726.1 | - | 1 | 220 | |||
BQ185626.1 | - | 1 | 31 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
