siRNA Details
Gene Symbol: PLEKHG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042663.1 | NP_001036128.1 | 20 | 2484 | ||
NM_001042664.1 | NP_001036129.1 | 19 | 2438 | |||
NM_001042665.1 | NP_001036130.1 | 19 | 2421 | |||
NM_001265592.1 | NP_001252521.1 | 20 | 2517 | |||
NM_001265593.1 | NP_001252522.1 | 19 | 2448 | |||
NM_001265594.1 | NP_001252523.1 | 19 | 2452 | |||
NM_020631.4 | NP_065682.2 | 19 | 2367 | |||
NM_198681.3 | NP_941374.2 | 20 | 2996 | |||
GenBank mRNA | AB018263.2 | - | 20 | 2472 | ||
AB097001.1 | - | 18 | 2207 | |||
AK024676.1 | - | 19 | 2366 | |||
AK091201.1 | - | 7 | 1417 | |||
AK096347.1 | - | 20 | 2944 | |||
AK125550.1 | - | 20 | 2450 | |||
AK131074.1 | - | 17 | 2691 | |||
AK294875.1 | - | 20 | 2517 | |||
AK299523.1 | - | 19 | 2448 | |||
AK301239.1 | - | 13 | 1447 | |||
BC015231.1 | - | 19 | 2428 | 3900065 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
