siRNA Details
Gene Symbol: PLEKHG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042663.1 | NP_001036128.1 | 9 | 1020 | ||
NM_001042664.1 | NP_001036129.1 | 8 | 974 | |||
NM_001042665.1 | NP_001036130.1 | 8 | 957 | |||
NM_001265592.1 | NP_001252521.1 | 9 | 1053 | |||
NM_001265593.1 | NP_001252522.1 | 8 | 984 | |||
NM_001265594.1 | NP_001252523.1 | 8 | 988 | |||
NM_020631.4 | NP_065682.2 | 8 | 903 | |||
NM_198681.3 | NP_941374.2 | 9 | 1532 | |||
GenBank mRNA | AB018263.2 | - | 9 | 1008 | ||
AB097001.1 | - | 7 | 749 | |||
AK024676.1 | - | 8 | 902 | |||
AK096347.1 | - | 9 | 1480 | |||
AK125550.1 | - | 9 | 986 | |||
AK131074.1 | - | 6 | 1227 | |||
AK294875.1 | - | 9 | 1053 | |||
AK299523.1 | - | 8 | 984 | |||
BC015231.1 | - | 8 | 964 | 3900065 | ||
BC036671.2 | - | 9 | 1533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
