siRNA Details
Gene Symbol: CCL15-CCL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027921.3 | 8 | 1435 | |||
NR_027922.3 | 7 | 1387 |
Gene Symbol: CCL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032962.4 | NP_116738.1 | 4 | 525 | ||
NM_032963.3 | NP_116739.1 | 3 | 477 | |||
GenBank mRNA | AK122606.1 | - | 2 | 1327 | ||
BC038289.1 | - | 3 | 442 | 5222914 | ||
BC045165.1 | - | 3 | 459 | 5225523 | ||
BI496964.1 | - | 1 | 133 | |||
DA821276.1 | - | 3 | 479 | |||
Z49270.1 | - | 3 | 452 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
